コンテンツへスキップ
Merck
Transmembrane redox sensor of ryanodine receptor complex.
The Journal of biological chemistry (2000-09-22)
W Feng, G Liu, P D Allen, I N Pessah
要旨
Inositol 1,4,5-trisphosphate receptors (IP(3)R) and ryanodine receptors (RyR) mediate the release of endoplasmic and sarcoplasmic reticulum (ER/SR) Ca(2+) stores and regulate Ca(2+) entry through voltage-dependent or ligand-gated channels of the plasma membrane. A prominent property of ER/SR Ca(2+) channels is exquisite sensitivity to sulfhydryl-modifying reagents. A plausible role for sulfhydryl chemistry in physiologic regulation of Ca(2+) release channels and the fidelity of Ca(2+) release from ER/SR is lacking. This study reveals the existence of a transmembrane redox sensor within the RyR1 channel complex that confers tight regulation of channel activity in response to changes in transmembrane redox potential produced by cytoplasmic and luminal glutathione. A transporter selective for glutathione is co-localized with RyR1 within the SR membrane to maintain local redox potential gradients consistent with redox regulation of ER/SR Ca(2+) release. Hyperreactive sulfhydryls previously shown to reside within the RyR1 complex (Liu, G., and Pessah, I. N. (1994) J. Biol. Chem. 269, 33028-33034) are an essential biochemical component of a transmembrane redox sensor. Transmembrane redox sensing may represent a fundamental mechanism by which ER/SR Ca(2+) channels respond to localized changes in transmembrane glutathione redox potential produced by physiologic and pathophysiologic modulators of Ca(2+) release from stores.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
7-ジメチルアミノ-3-(4-マレイミドフェニル)-4-メチルクマリン, suitable for fluorescence, BioReagent, ≥90% (HPCE)
Sigma-Aldrich
7-ジメチルアミノ-3-(4-マレイミドフェニル)-4-メチルクマリン, ≥95% (HPLC), solid