コンテンツへスキップ
Merck
  • Vegetables and PUFA-rich plant oil reduce DNA strand breaks in individuals with type 2 diabetes.
Vegetables and PUFA-rich plant oil reduce DNA strand breaks in individuals with type 2 diabetes.
Molecular nutrition & food research (2012-11-14)
Elisabeth Müllner, Helmut Brath, Simone Pleifer, Christiane Schiermayr, Andreas Baierl, Marlies Wallner, Theresia Fastian, Yvonne Millner, Kristina Paller, Trine Henriksen, Henrik Enghusen Poulsen, Ernst Forster, Karl-Heinz Wagner
要旨
Type 2 diabetes is a multifactorial disease associated with increased oxidative stress, which may lead to increased DNA damage. The aim of this study was to investigate the effect of a healthy diet on DNA oxidation in diabetics and nondiabetics. Seventy-six diabetic and 21 nondiabetic individuals participated in this study. All subjects received information about the benefits of a healthy diet, while subjects randomly assigned to the intervention group received additionally 300 g of vegetables and 25 mL PUFA-rich plant oil per day. DNA damage in mononuclear cells (Comet Assay), urinary excretion of 8-oxo-7-hydro-2'-deoxyguanosine (8-oxodG) and 8-oxo-7,8-dihydroguanosine (8-oxoGuo) and glycated hemoglobin (HbA1c) were measured at baseline, after 4, 8 (end of intervention), and 16 weeks. The intervention with vegetables and PUFA-rich oil led to a significant increase in plasma antioxidant concentrations. Diabetic individuals of the intervention group showed a significant reduction in HbA1c and DNA strand breaks. Levels of HbA1c were also improved in diabetics of the information group, but oxidative damage to DNA was not altered. Urinary 8-oxodG and 8-oxoGuo excretion remained unchanged in both groups. This study provides evidence that a healthy diet rich in antioxidants reduces levels of DNA strand breaks in diabetic individuals.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
8-ヒドロキシ-2′-デオキシグアノシン, ≥98% (TLC)