コンテンツへスキップ
Merck
  • Homoplantaginin Inhibits Palmitic Acid-induced Endothelial Cells Inflammation by Suppressing TLR4 and NLRP3 Inflammasome.
Homoplantaginin Inhibits Palmitic Acid-induced Endothelial Cells Inflammation by Suppressing TLR4 and NLRP3 Inflammasome.
Journal of cardiovascular pharmacology (2015-09-12)
Baiqiu He, Baobao Zhang, Feihua Wu, Liying Wang, Xiaoji Shi, Weiwei Qin, Yining Lin, Shiping Ma, Jingyu Liang
要旨
Palmitic acid (PA)-induced vascular endothelial inflammation plays a pivotal role in the occurrence and development of vascular diseases. The present study was conducted to examine the effect of homoplantaginin, a main flavonoid from a traditional Chinese medicine Salvia plebeia R. Br., on PA-treated human umbilical vein endothelial cells inflammation and the underlying molecular mechanism. Firstly, we found that homoplantaginin (0.1, 1, 10 μM) dose-dependently reduced expression of toll-like receptor-4 evoked by PA (100 μM). The inhibitory effect of homoplantaginin was further confirmed under lipopolysaccharide challenge. In addition, downstream adapted proteins including myeloid differentiation primary response gene 88, toll/interleukin-1 receptor-domain containing adaptor-inducing interferon-β and tumor necrosis factors receptor associated factor-6 were successfully inhibited by homoplantaginin under PA treatment. Also, we found that homoplantaginin tightly controlled PA-induced reactive oxygen species to prevent nucleotide-binding domain-like receptor 3 (NLRP3) inflammasome activation by suppressing reactive oxygen species-sensitive thioredoxin-interacting protein, NLRP3, and caspase-1. Meanwhile, protein and mRNA levels of inflammatory mediators (interleukin-1β, intercellular cell adhesion molecule-1, and monocyte chemotactic protein-1) were decreased by homoplantaginin. Furthermore, homoplantaginin restored PA-impaired nitric oxide generation. Taken together, these results indicated that homoplantaginin protected endothelial cells from ameliorating PA-induced endothelial inflammation via suppressing toll-like receptor-4 and NLRP3 pathways, and restoring nitric oxide generation, suggesting it may be a potential candidate for further development in the prevention and treatment of vascular diseases.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
Homoplantaginin, ≥98% (HPLC)