ステロイドの分析、Ascentis Express F5、アセトニトリル移動相
材料
分析カラム
column
Ascentis Express F5, 10 cm x 2.1 mm I.D., 2.7 μm particles (53569-U)
mobile phase
[A] water; [B] acetonitrile; (70:30, A:B)
flow rate
0.2 mL/min
column temp.
35 °C
detector
UV, 254 nm
injection
2 μL
sample
50 mg/L in 75:25, water:methanol