Merck
HPLCによる2-ブロモ-3-メチル酪酸エナンチオマーの分析、CHIROBIOTIC® R
HPLCによる2-ブロモ-3-メチル酪酸エナンチオマーの分析、CHIROBIOTIC® R application for HPLC
材料
Standard
製品番号
詳細
価格
(S)-(−)-2-Bromo-3-methylbutyric acid
96%
2-ブロモ-3-メチル酪酸
97%
(R)-(+)-2-Bromo-3-methylbutyric acid
96%
分析カラム
製品番号
詳細
価格
Astec®CHIROBIOTIC®RキラルHPLCカラム
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC R, 25 cm x 4.6 mm I.D., 5 μm particles (13024AST)
mobile phase
[A] 20 mM ammonium acetate; [B] methanol; (15:85, A:B)
(alternate mobile phase of 0.1% ammonium acetate in methanol gives 3.5 and 4.3 min retention)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
5 μL
sample
2 mg/mL in methanol