Merck
HPLCによるアフラトキシンの分析、Ascentis®Express C18
HPLCによるアフラトキシンの分析、Ascentis®Express C18 application for HPLC
材料
Standard
製品番号
詳細
価格
アフラトキシンMix 4成分 溶液
analytical standard
分析カラム
製品番号
詳細
価格
Ascentis® Express C18, 2.7 µm HPLCカラム
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
CONDITIONS
column
Ascentis Express C18, 10 cm x 2.1 mm I.D., 2.7 μm particles (53823-U)
mobile phase
[A] water; [B] methanol; [C] acetonitrile; (720:120:120, A:B:C) containing 0.780 g/L potassium bromide, 230 μL/L nitric acid
flow rate
0.400 mL/min
pressure
4568 psi (315 bar)
column temp.
35 °C
detector
FLD excitation = 360 nm, emission = 440 nm
injection
20 μL
sample
200 μL spiked 84:16 acetonitrile:di water in 880 μL di water, Spike concentrations: B1 = 16 ppb; B2 = 4 ppb; G1 = 16 ppb; B2 = 4 ppb
詳細
アナリシスノート
Aflatoxins are a complex of closely related toxins produced as secondary metablolites of fungi. They are found in grains and nuts such as corn, wheat, or peanuts. They are produced by aspergillus flavus and a. Parasiticus. Members of the group include aflatoxin b1, aflatoxin b2, aflatoxin g1, aflatoxin g2, aflatoxin m1, and aflatoxin m2.