HPLCによるフルルビプロフェンエナンチオマーの分析、Astec® Cellulose DMP
CONDITIONS
column
Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase
[A] heptane; [B] isopropanol; [C] trifluoroacetic acid (95:5:0.1, A:B:C)
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
3 μL
sample
1 g/L in methanol