HPLCによるケトロラクエナンチオマーの分析、CHIROBIOTIC® T
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
0.1 wt% mM ammonium acetate in methanol
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 230 nm