콘텐츠로 건너뛰기
Merck
  • A new reactivity pattern for vinyl bromides: cine-substitution via palladium catalysed C-N coupling/Michael addition reactions.
A new reactivity pattern for vinyl bromides: cine-substitution via palladium catalysed C-N coupling/Michael addition reactions.
Organic & biomolecular chemistry (2005-08-18)
Michael C Willis, Jay Chauhan, William G Whittingham
초록
Palladium catalysed C-N bond formation can be used to convert vinyl bromides to the corresponding enamines, which are reacted in situ with alkylidene malonates to provide Michael adducts. The overall transformation results in cine-substitution of the starting vinyl bromide.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Vinyl bromide, 98%
Sigma-Aldrich
Vinyl bromide solution, 1.0 M in THF