콘텐츠로 건너뛰기
Merck
  • Inhibition of amyloid fibrillation of lysozyme by indole derivatives--possible mechanism of action.
Inhibition of amyloid fibrillation of lysozyme by indole derivatives--possible mechanism of action.
The FEBS journal (2007-11-22)
Dina Morshedi, Nasrollah Rezaei-Ghaleh, Azadeh Ebrahim-Habibi, Shahin Ahmadian, Mohsen Nemat-Gorgani
초록
Amyloid aggregation of polypeptides is related to a growing number of pathologic states known as amyloid disorders. There is a great deal of interest in developing small molecule inhibitors of the amyloidogenic processes. In the present article, the inhibitory effects of some indole derivatives on amyloid fibrillation of hen egg white lysozyme (HEWL) are reported. Acidic pH and high temperatures were used to drive HEWL towards amyloid formation. A variety of techniques, ranging from thioflavin T fluorescence and Congo red absorbance assays to far-UV CD and transmission electron microscopy, were employed to characterize the HEWL fibrillation process. Among the indole derivatives tested, indole 3-acetic acid, indole 3-carbinol and tryptophol had the most inhibitory effects on amyloid formation, indole and indole 3-propionic acid gave some inhibition, and indole aldehyde and tryptophan showed no significant inhibition. Although indoles did not protect the HEWL native state from conformational changes, they were effective in diminishing HEWL amyloid fibril formation, delaying both the nucleation and elongation phases. Disaggregation of previously formed HEWL amyloid fibrils was also enhanced by indole 3-acetic acid. Various medium conditions, such as the presence of different anions and alcoholic cosolvents, were explored to gain an insight into possible mechanisms. These observations, taken together, suggest that the indole ring is likely to play the main role in inhibition and that the side chain hydroxyl group may contribute positively, in contrast to the side chain carbonyl and intervening methylene groups.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
3-(2-Hydroxyethyl)indole, 97%