콘텐츠로 건너뛰기
Merck
  • Modifications around the hydroxamic acid chelating group of fosmidomycin, an inhibitor of the metalloenzyme 1-deoxyxylulose 5-phosphate reductoisomerase (DXR).
Modifications around the hydroxamic acid chelating group of fosmidomycin, an inhibitor of the metalloenzyme 1-deoxyxylulose 5-phosphate reductoisomerase (DXR).
Bioorganic & medicinal chemistry letters (2012-10-03)
Catherine Zinglé, Lionel Kuntz, Denis Tritsch, Catherine Grosdemange-Billiard, Michel Rohmer
초록
Fosmidomycin derivatives in which the hydroxamic acid group has been replaced by several bidentate chelators as potential hydroxamic alternatives were prepared and tested against the DXR from Escherichia coli. These results illustrate the predominant role of the hydroxamate functional group as the most effective metal binding group in DXR inhibitors.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Fosmidomycin sodium salt hydrate, ≥95% (NMR)