MilliporeSigma
Intensified and Continuous Downstream Processing
Gain operational efficiency and flexibility while reducing footprint and costs
HomeIntegrated OfferingsBiopharma 4.0Intensified & Continuous Downstream ProcessingUltrafiltration Platform
Looking to increase your downstream process efficiency?
Transformation of the biomanufacturing workflow is being accelerated by the demand for new therapeutics combined with the need to reduce costs while ensuring operational flexibility and productivity.
Process intensification, closed, and continuous bioprocessing are foundational strategies for achieving these goals. They are widely and successfully deployed in the downstream suite to be more efficient, faster, smaller, less expensive, and sustainable.
To help biopharmaceutical manufacturers intensify, closing, digitally-enhance, or go fully continuous with their processes and gain new levels of operational efficiency and flexibility, there are multiple levers within the DSP suite.
Ultrafiltration
Use of single-pass tangential flow filtration (SPTFF) eliminates the need for hold tanks or a retentate loop, allowing flow-through operation and intensified downstream processing of monoclonal antibodies. Productivity and efficiency of the mAb DSP workflow can be further increased by coupling SPTFF with chromatography unit operations. SPTFF is also uniquely positioned to enable high-concentration formulations with improved product recovery during the final formulation step.
  •  Pellicon® SPTFF offers an unparalleled solution to reduce in-process volumes within downstream processing of drug products, reach concentrations and yields unachievable with batch TFF due to working volume limits, and enable connected and continuous processes.
  • Pellicon® Cassettes consistently and reliably deliver purity assurance at every stage and scale throughout the life of a drug product.
  • Pellicon® Capsules for the filtration of feedstreams that require single-use capabilities, including enhanced ease-of-use, process flexibility, product turnaround, and reduced operator exposure.
Related Categories
Operator, fully gowned, standing in front of the Mobius Multi Column Capture System
Chromatography Systems for Bioprocessing
Single-use and multi-use chromatography systems are critical for the separation of your valuable molecules.
4 different sizes of Affinity chromatography with Eshmuno® resin bottles sit on a black counter with a scientist working on a piece of machinery in the background
Affinity Chromatography Resins
Affinity chromatography is often used during the capture phase to isolate the target molecule and reduce process volume.
4 different sizes of Eshmuno CMX, ion exchange chromatography resin bottles
Ion Exchange Chromatography Resins
Ion exchange (IEX) chromatography separates biomolecules based on difference in charge between the target molecule and chromatography resin.
3 sizes of Natrix Q chromatography membrane devices
Membrane Chromatography
Optimize downstream purification using membrane chromatography for capture and polishing. Natrix membranes boost economics and facility efficiency.
Pelicon cassette lab-scale single-pass tangential flow filtration setup
Tangential Flow Filtration Devices
Elevate Filtration Precision: Our tangential flow products ensure purity for mAbs, vaccines, and more, scaling from single-use to multi-use.
Related Resources
Related Webinars
Effective Implementation of Multi Column Chromatography
Improve your process efficiency and reduce costs
Related Videos
BioContinuum™ Platform: Your enabler of the biomanufacturing facility of the future!