Skip to Content
Merck
All Photos(1)

Key Documents

49281

Millipore

Glucose Salt Teepol Broth

suitable for microbiology

Synonym(s):

GSTB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

form

solid

shelf life

limited shelf life, expiry date on the label

composition

glucose, 5 g/L
meat extract, 3 g/L
methyl violet, 0.002 g/L
peptone, 10 g/L
sodium chloride, 30 g/L

final pH

8.6±0.2 (25 °C)

application(s)

environmental
food and beverages

microbiology

suitability

selective for Vibrio spp.

Application

For selective enrichment and enumeration (MPN method) of Vibrio parahemolyticus from food products.

Legal Information

Teepol is a trademark of Shell Chemical Co.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Sakazaki et al.
Journal of Food Protection, 49, 773-773 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service