MilliporeSigma
ASTS-012
Air Science USA LLC
Air Science Filters
AMM Carbon Filter
Shipped & Sold By Air Science
A Trusted Marketplace Seller
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
General description
Following filters are available for Ductless Fume Hoods: ASTS-001 GP Plus! Carbon Filter for general purpose for organics, solvents, alcohols, hydrocarbons, and odors, ASTS-015 ACI Plus! Carbon Filter for acids, ASTS-013 FOR Carbon Filter for aldehydes, ASTS-012 AMM Carbon Filter for ammonia, ASTS-030 HEPA Filter for powders and particulates, ASTS-PRF 6-Pack Prefilters for Dust and to extend Main Filter Life.
Legal Information
Purair is a registered trademark of Air Science USA LLC
Still not finding the right product?
Explore all of our products under Air Science Filters
Choose from one of the most recent versions:
Certificates of Analysis (COA)