Skip to Content
Merck
All Photos(1)

Key Documents

160679

Sigma-Aldrich

2-Chlorostyrene

97%, contains 0.1% hydroquinone as stabilizer

Synonym(s):

1-Chloro-2-ethenylbenzene, 1-Chloro-2-vinylbenzene, o -Chlorostyrene, o -Chlorovinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC6H4Cl
CAS Number:
Molecular Weight:
138.59
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Pricing and availability is not currently available.

assay

97%

form

liquid

contains

0.1% hydroquinone as stabilizer

refractive index

n20/D 1.565 (lit.)

bp

58-60 °C/7 mmHg (lit.)

density

1.08 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Clc1ccccc1C=C

InChI

1S/C8H7Cl/c1-2-7-5-3-4-6-8(7)9/h2-6H,1H2

InChI key

ISRGONDNXBCDBM-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Carc. 1B - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

138.2 °F - closed cup

flash_point_c

59 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leire Ruiz-Rubio et al.
Materials (Basel, Switzerland), 12(1) (2019-01-10)
Polymeric thin films patterned with honeycomb structures were prepared from poly(x-chlorostyrene) and statistical poly(x-chlorostyrene-co-styrene) copolymers by static breath figure method. Each polymeric sample was synthesized by free radical polymerization and its solution in tetrahydrofuran cast on glass wafers under 90%

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service