Skip to Content
Merck
All Photos(1)

Key Documents

389919

Sigma-Aldrich

3,3′-Diethylthiatricarbocyanine perchlorate

99%

Synonym(s):

DTTC perchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H25ClN2O4S2
CAS Number:
Molecular Weight:
517.06
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

99%

mp

184 °C (dec.) (lit.)

λmax

760 nm

SMILES string

[O-]Cl(=O)(=O)=O.CCN1\C(Sc2ccccc12)=C\C=C\C=C\C=C\c3sc4ccccc4[n+]3CC

InChI

1S/C25H25N2S2.ClHO4/c1-3-26-20-14-10-12-16-22(20)28-24(26)18-8-6-5-7-9-19-25-27(4-2)21-15-11-13-17-23(21)29-25;2-1(3,4)5/h5-19H,3-4H2,1-2H3;(H,2,3,4,5)/q+1;/p-1

InChI key

PZQHZVAROUTGBU-UHFFFAOYSA-M

Application

Efficient laser dye for pulsed operations

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changning Wang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(7), 2382-2390 (2011-02-18)
Myelination is one of the fundamental biological processes in the development of vertebrate nervous system. Disturbance of myelination is found to be associated with progression in many neurological diseases such as multiple sclerosis. Tremendous efforts have been made to develop
B Stieger et al.
Biochimica et biophysica acta, 732(1), 324-326 (1983-07-13)
The sensitivity of the fluorescent dye, 3,3'-diethylthiadicarbocyanine (DiS-C2(5], was too low for the detection of membrane potential changes in rat small intestinal membrane vesicles. Only after adding LaCl3 or after fractionation of the intestinal membranes by free-flow electrophoresis could the
K E Akerman et al.
Journal of neurochemistry, 48(2), 552-559 (1987-02-01)
Membrane potentials of particles present in a subcellular brain preparation, called synaptoneurosomes, have been monitored by measurement of changes in the absorbance of a cyanine dye, DiS-C2-5. The membrane potential of the particles seems to be dependent on both Cl-
M O Enkvist et al.
Brain research, 462(1), 67-75 (1988-10-11)
The cyanine dye DiS-C2-(5) was used to investigate the effect of K+ and glutamate receptor agonists on the membrane potential of whole populations of primary rat astrocytes in suspension. Increasing the external K+ concentration from 5 to 40 mM caused
Eddy Kuwana et al.
Biophysical journal, 83(2), 1165-1176 (2002-07-19)
To investigate fluorescence lifetime spectroscopy in tissue-like scattering, measurements of phase modulation as a function of modulation frequency were made using two fluorescent dyes exhibiting single exponential decay kinetics in a 2% intralipid solution. To experimentally simulate fluorescence multiexponential decay

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service