Skip to Content
Merck
All Photos(1)

Key Documents

591866

Sigma-Aldrich

Poly(2,2,2-trifluoroethyl methacrylate-co-glycidyl methacrylate)

glycidyl methacrylate 10 mol %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H7F3O2)m(C7H10O3)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Pricing and availability is not currently available.

form

solid

composition

glycidyl methacrylate, 10 mol %

viscosity

10.25 cP, 1 % in methylene chloride

mp

52-58 °C (lit.)

solubility

DMSO: soluble
NMP: soluble
THF: soluble
methylene chloride: soluble

Looking for similar products? Visit Product Comparison Guide

General description

Transparent, low surface tension polymer.

Application

Low refractive index, photocrosslinkable polymer for cladding layer in optical waveguides.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service