Skip to Content
Merck
All Photos(1)

Key Documents

630098

Sigma-Aldrich

Poly(2,2,2-trifluoroethyl acrylate)

Synonym(s):

2,2,2-Trifluoroethyl acrylate polymer, Polytrifluoroethyl acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CO2CH2CF3)]n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

refractive index

n20/D 1.411 (lit.)

transition temp

Tg -10 °C(lit.)

density

1.403 g/mL at 25 °C (lit.)

Application

Low refractive index polymer for cladding layer in optical waveguides. Transparent, low surface tension polymer.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service