Skip to Content
Merck
All Photos(1)

Key Documents

630152

Sigma-Aldrich

Poly(1,1,1,3,3,3-hexafluoroisopropyl acrylate)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
(C6H4F6O2)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

refractive index

n20/D 1.375 (lit.)

transition temp

Tg -23 °C

density

1.562 g/mL at 25 °C (lit.)

Application

Low refractive index polymer for cladding layer in optical waveguides.
Transparent, low surface tension polymer.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service