Skip to Content
Merck
All Photos(4)

Key Documents

806188

Sigma-Aldrich

Diethylammonium iodide

greener alternative

Synonym(s):

Diethylamine Hydroiodide, Greatcell Solar®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H12IN
CAS Number:
Molecular Weight:
201.05
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.23

Pricing and availability is not currently available.

assay

98%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

301.6 °C

greener alternative category

SMILES string

CCNCC.I

InChI

1S/C4H11N.HI/c1-3-5-4-2;/h5H,3-4H2,1-2H3;1H

InChI key

YYMLRIWBISZOMT-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Diethylammonium iodide (DAI), a two dimensional (2D) organic-inorganic halide material that is used in the crystallization of perovskites, improves the quality of the films. Its properties include moisture resistance and hydrophobicity that make it useful in the formation of perovskite films. It further results in the fabrication of perovskite-based solar cells with a high absorption coefficient and good power conversion efficiency (PCE).[1][2]
The iodide and bromide based alkylated halides find applications as precursors for fabrication of perovskites for photovoltaic applications.[3][4][5]

Legal Information

Product of Greatcell Solar®
Greatcell Solar is a registered trademark of Greatcell Solar

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced efficiency and light stability of planar perovskite solar cells by diethylammonium bromide induced large-grain 2D/3D hybrid film
Huang X, et al.
Organic Electronics, 67, 101-108 (2019)
Two-dimensional additive diethylammonium iodide promoting crystal growth for efficient and stable perovskite solar cells
Huang X, et al.
Royal Society of Chemistry Advances, 9(14), 7984-7991 (2019)
Nam Joong Jeon et al.
Nature, 517(7535), 476-480 (2015-01-07)
Of the many materials and methodologies aimed at producing low-cost, efficient photovoltaic cells, inorganic-organic lead halide perovskite materials appear particularly promising for next-generation solar devices owing to their high power conversion efficiency. The highest efficiencies reported for perovskite solar cells
Wei Zhang et al.
Nano letters, 15(3), 1698-1702 (2015-02-05)
The performance of perovskite solar cells has been progressing over the past few years and efficiency is likely to continue to increase. However, a negative aspect for the integration of perovskite solar cells in the built environment is that the
Zhi-Kuang Tan et al.
Nature nanotechnology, 9(9), 687-692 (2014-08-05)
Solid-state light-emitting devices based on direct-bandgap semiconductors have, over the past two decades, been utilized as energy-efficient sources of lighting. However, fabrication of these devices typically relies on expensive high-temperature and high-vacuum processes, rendering them uneconomical for use in large-area

Articles

Next generation solar cells have the potential to achieve conversion efficiencies beyond the Shockley-Queisser (S-Q) limit while also significantly lowering production costs.

Dr. Perini and Professor Correa-Baena discuss the latest research and effort to obtain higher performance and stability of perovskite materials.

For several decades, the need for an environmentally sustainable and commercially viable source of energy has driven extensive research aimed at achieving high efficiency power generation systems that can be manufactured at low cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service