901933
NORIT® SX Plus F CAT
Synonym(s):
Activated carbon, Activated charcoal
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
Methylene blue adsorption 20g/100 g
bulk density
200‑300 kg/m3
InChI
1S/C
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Application
Activated carbons are internally porous microcrystalline, non-graphitic forms of carbon. The activated carbons possess large surface area (~1000 m2/g) and pore volumes which make them useful for applications in energy storage devices, catalysis, and for removal of impurities from gases and liquids.
NORIT® SX PLUS F CAT is a powdered activated carbon having broad particles size distribution. It is designed for use as carrier of precious metals in liquid phase pharmaceutical and fine chemical synthesis. Its unique raw material enhances the metal dispersion over its surface and thereby reduces the need of metal to achieve the same catalytic activity.
The high porosity of NORIT® SX PLUS F CAT results in a high adsorption capacity for a large range of impurities originating from both the reactants and byproducts. Adsorption of impurities improves the purity of the end product and overcomes the issue of catalyst poisoning and leads to longer catalyst life.
NORIT® SX PLUS F CAT is a powdered activated carbon having broad particles size distribution. It is designed for use as carrier of precious metals in liquid phase pharmaceutical and fine chemical synthesis. Its unique raw material enhances the metal dispersion over its surface and thereby reduces the need of metal to achieve the same catalytic activity.
The high porosity of NORIT® SX PLUS F CAT results in a high adsorption capacity for a large range of impurities originating from both the reactants and byproducts. Adsorption of impurities improves the purity of the end product and overcomes the issue of catalyst poisoning and leads to longer catalyst life.
Legal Information
Product of Cabot Corporation.
NORIT® is a registered trademark of Cabot Corporation.
NORIT® is a registered trademark of Cabot Corporation.
Norit is a registered trademark of Norit N.V.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Activated Carbon for Water and Wastewater treatment
Cecen F and Aktas O
Wiley-VCH (2011)
Energy storage applications of activated carbons: supercapacitors and hydrogen storage.
Sevilla M and Mokaya R
Energy & Environmental Science, 7, 1250-1280 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service