Skip to Content
Merck
All Photos(2)

Key Documents

901939

Sigma-Aldrich

NORIT® RX1.5 EXTRA

Synonym(s):

Activated carbon, Activated charcoal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C
CAS Number:
UNSPSC Code:
23201100
NACRES:
NA.23

description

ash content: 3 mass %

form

powder

hardness

99 (Ball-pan hardness)

bulk density

350‑450 kg/m3

anion traces

chloride (Cl-): 0.2 mass% (acid extr.)

cation traces

Cu: 8 mg/kg (acid extr.)
Zn: 2 mg/kg (acid extr.)

InChI

1S/C

Inchi Key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Application

Activated carbons are internally porous microcrystalline, non-graphitic forms of carbon. The activated carbons possess large surface area (~1000 m2/g) and pore volumes which make them useful for applications in energy storage devices, catalysis, and for removal of impurities from gases and liquids.

NORIT® RX 1.5 EXTRA is an acid washed steam activated carbon which possesses high pore volume. Its excellent purity along with good abrasion resistance makes it suitable as a precious metal catalyst carrier.

Legal Information

Product of Cabot Corporation.
NORIT® is a registered trademark of Cabot Corporation.
Norit is a registered trademark of Norit N.V.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activated Carbon for Water and Wastewater treatment
Cecen F and Aktas O
Wiley-VCH (2011)
Energy storage applications of activated carbons: supercapacitors and hydrogen storage.
Sevilla M and Mokaya R
Energy & Environmental Science, 7, 1250-1280 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service