Skip to Content
Merck
All Photos(1)

Key Documents

925012

Sigma-Aldrich

E7820-PEG5-NH2 hydrochloride

Synonym(s):

N-(17-amino-3,6,9,12,15-pentaoxaheptadecyl)-3-(N-(3-cyano-4-methyl-1H-indol-7-yl)sulfamoyl)benzamide hydrochloride, Crosslinker−E3 Ligase ligand conjugate, DCAF15 protein degrader building block

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H39N5O8S · xHCl
Molecular Weight:
617.71 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.22

ligand

E7820

Quality Level

reaction suitability

reactivity: carboxyl reactive
reagent type: ligand-linker conjugate

functional group

amine

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Protein degrader builiding block E7820-PEG5-NH2 hydrochloride enables the synthesis of molecules for targeted protein degradation and PROTAC® (proteolysis-targeting chimeras) research. This conjugate contains a DCAF15-recruiting ligand E7820 (SML2950), a PEG linker, and a pendant amine for reactivity with a carboxylic acid on the target ligand. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and degrader, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks with a terminal amine, parallel synthesis can be used to more quickly generate degrader libraries that feature variation in crosslinker length, composition, and E3 ligase ligand.

Technology Spotlight: DCAF15 Degrader Building Blocks for Targeted Protein Degradation

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service