Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

939447

Sigma-Aldrich

Dextran aldehyde

new

40% degree of functionalization

Synonym(s):

Aldehyde functionalized dextran, DexOx, Dextran-CHO, Oxidized dextran

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H18O10)n
UNSPSC Code:
51131911

Quality Level

description

natural polymers

form

powder or chunks (or Fibers)

color

white to light yellow-brown

functional group

aldehyde

storage temp.

2-8°C

General description

Dextran aldehyde, or aldehyde functionalized dextran, is a versatile, high quality natural polymer used in a variety of biomedical research applications. Dextran, a naturally synthesized polysaccharide, offers excellent biocompatibility, water solubility and low toxicity, minimizing any adverse effects on cellular behavior. The aldehyde function groups present on the dextran backbone can react with amine or hydrazide groups present on a variety of biomolecules or other polymers.

Application

Applications of dextran aldehyde include 3D cell culture and cell encapsulation, tissue engineering, wound healing, tissue adhesives, drug delivery, and anti-adhesion barriers on medical devices. It is also used as a biocatalyst and the chemical engineering enzymes. The aldehyde functional group enable efficient crosslinking with a variety amine containing biomolecules, including proteins, peptides, antibodies, and enzymes. It can also crosslink with other natural or synthetic polymers to form biocompatible hydrogels, drug delivery systems, or other tissue engineering scaffolds.

Features and Benefits

  • Biomedical research grade- Our dextran aldehyde is produced with the utmost quality control to ensure reliable performance.
  • Versatile applications – Dextran aldehyde can be utilized in a wide range of biomedical research areas.
  • Enhanced biocompatibility- Dextran aldehyde is specifically designed to maintain excellent biocompatibility for use in biomedical research.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stabilization of Multimeric Enzymes via Immobilization and Further Cross-Linking with Aldehyde-Dextran
Mateo C, et al.
Methods in Molecular Biology, 175?187-175?187 (2020)
Dextran Aldehyde in Biocatalysis: More Than a Mere Immobilization System
Tacias-Pascacio G V, et al.
Catalysts, 9(7), 622-622 (2019)
Rational construction of a robust nanoprobe for highly selective and sensitive nitrite and formaldehyde detection and imaging in real foods
Tan L, et al.
Food Chemistry, 405, 134949-134949 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service