GF04956598
Calcium
lump, 10 mm max. lump size, weight 200 g, purity 99%
Synonym(s):
CA006101
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
assay
99%
form
lumps
manufacturer/tradename
Goodfellow 049-565-98
weight
200 g
max. lump size
10 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
signalword
Danger
hcodes
pcodes
Hazard Classifications
Water-react 2
Storage Class
4.3 - Hazardous materials which set free flammable gases upon contact with water
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service