Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF67647347

Lead

foil, 10mm disks, thickness 0.0125mm, 99.99+%

Synonym(s):

Lead, PB000130

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€75.00
1 ML
€422.00

€75.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€75.00
1 ML
€422.00

About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12141723
PubChem Substance ID:
NACRES:
NA.23

€75.00


Please contact Customer Service for Availability

Request a Bulk Order

assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 676-473-47

resistivity

20.65 μΩ-cm

diam. × thickness

10 mm × 0.0125 mm

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF05860739GF30031546GF40522520
Lead foil, 10mm disks, thickness 0.0125mm, 99.99+%

GF67647347

Lead

Lead foil, 10mm disks, thickness 0.025mm, 99.99+%

GF05860739

Lead

Lead foil, 10mm disks, thickness 0.007mm, 99.99+%

GF30031546

Lead

Lead foil, 10mm disks, thickness 0.04mm, 99.99+%

GF40522520

Lead

form

foil

form

foil

form

foil

form

foil

manufacturer/tradename

Goodfellow 676-473-47

manufacturer/tradename

Goodfellow 058-607-39

manufacturer/tradename

Goodfellow 300-315-46

manufacturer/tradename

Goodfellow 405-225-20

bp

1740 °C (lit.)

bp

1740 °C (lit.)

bp

1740 °C (lit.)

bp

1740 °C (lit.)

resistivity

20.65 μΩ-cm

resistivity

20.65 μΩ-cm

resistivity

20.65 μΩ-cm

resistivity

20.65 μΩ-cm

mp

327.4 °C (lit.)

mp

327.4 °C (lit.)

mp

327.4 °C (lit.)

mp

327.4 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

pictograms

Health hazardEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

target_organs

Central nervous system,Blood,Immune system,Kidney

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    D W Rogers
    Medical physics, 26(4), 533-537 (1999-05-05)
    Data are presented to allow the photon beam quality specifier being used in the new AAPM TG-51 protocol, %dd(10)x, to be extracted from depth-dose data measured with a 1 mm lead foil either 50 cm or 30 cm from the
    I Sewerin
    Tandlaegebladet, 94(12), 483-486 (1990-09-01)
    The lead foil of dental x-ray film packets may cause a number of artifacts; especially well known are the "tractor treads" and "herringbone patterns" following a backward filmpositioning in the patient's mouth. In the present paper a specific and seldomly
    X A Li et al.
    Medical physics, 26(7), 1245-1251 (1999-08-06)
    The purpose of this work is to study the dose enhancement by a thin foil (thickness of 0.2-4 mm) of high-Z material in a water phantom, irradiated by high-energy photon beams. EGS4 Monte Carlo technique was used. Perturbations on the
    A Janhom et al.
    Dento maxillo facial radiology, 29(1), 20-27 (2000-02-02)
    To determine the interaction between image noise and file compression, with special emphasis on the accuracy of caries diagnosis. Fifty-nine bitewing radiographs of patients were taken simultaneously with Ektaspeed Plus (Eastman-Kodak, Rochester, NY, USA) film without lead foil and the
    H Fujita et al.
    The British journal of radiology, 82(978), 504-508 (2009-01-21)
    It is useful to utilise a computed radiography system with a combination of storage phosphor plates and therapy cassettes for portal images. However, the therapy cassette has some disadvantages. The objective of the present study was to determine whether the

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service