Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

700362O

Avanti

(E,E,E)-Geranylgeraniol

Avanti Research - A Croda Brand

Synonym(s):

(2E,6E,10E)-3,7,11,15-tetramethylhexadeca-2,6,10,14-tetraen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O
Molecular Weight:
290.48
MDL number:
UNSPSC Code:
12352212
NACRES:
NA.21

assay

>99% (TLC)

form

oil

packaging

pkg of 1 x 10 mg

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC\C=C(\CC\C=C(\CC\C=C(\CCC=C(C)C)/C)/C)/C

InChI

1S/C20H34O/c1-17(2)9-6-10-18(3)11-7-12-19(4)13-8-14-20(5)15-16-21/h9,11,13,15,21H,6-8,10,12,14,16H2,1-5H3/b18-11+,19-13+,20-15+

InChI key

OJISWRZIEWCUBN-QIRCYJPOSA-N

Packaging

5 mL Amber Glass Screw Cap Vial (700362O-10MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service