03-0197-00
SMC® Human SARS-CoV-2 S1 IgA Kit
detects antigen specific IgA in human serum and plasma at much lower levels compared to traditional immunoassays
About This Item
Recommended Products
manufacturer/tradename
SMC®
technique(s)
single molecule counting: suitable
assay range
inter-assay cv: <15%
intra-assay cv: <9%
(kit were studied using fourteen plasma samples, run in triplicate in three separate runs.)
General description
Application
Packaging
Components
- Assay Buffer
- SARS-CoV-2 S1 IgA Beads
- Standard Diluent
- SARS-CoV-2 S1 IgA Detection Antibody
- 10X Wash Buffer
- Buffer D
- Elution Buffer B
Other Notes
- For use with the state of the art SMCxPRO® Ultrasensitive Immunoassay System
- Qualitative assay; no standard curve
- For Research Use Only. Not For Use In Diagnostic Procedures
- Detects SARS-CoV-2 S1 specific IgA antibody as early as 3 days after PCR positive result
- Requires as little as 2μL of human serum and plasma sample (1:50 recommended dilution)
- Fast assay protocol (one hour capture, 30 minute detection steps)
Inflammation & Immunology, Immune Response, Infectious Diseases, Isotyping
Research Sub-Category:
Inflammatory Disease, Immune Response, Virology, Infectious Diseases - Viral, Immunoglobulins & Immunology
Legal Information
Disclaimer
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service