Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

05-1113

Sigma-Aldrich

Anti-Harmartin (TSC1) Antibody

ascites fluid, from mouse

Synonym(s):

Tuberous sclerosis 1 protein, tuberous sclerosis 1, tumor suppressor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TSC1(7248)

General description

Harmartin is the gene product of tuberous sclerosis-1 (TSC1). It is thought to have a tumor suppression function and also augments the expression of the TSC2 product tuberin by inhibiting its ubiquitination.

Specificity

This antibody recognizes Harmartin

Immunogen

Recombinant protein from human Harmartin

Application

Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected Harmartin in human skin tissue, melanocytes.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling
This Anti-Harmartin (TSC1) Antibody is validated for use in WB, IH(P) for the detection of Harmartin (TSC1).

Quality

Evaluated by Western Blot in NIH/3T3 cell lysate.
Western Blot Analysis: 1:500 dilution of this antibody detected Harmartin in 10 µg of NIH/3T3 cell lysate.

Target description

Approx. 160 kDa

Linkage

Replaces: 04-426

Physical form

Ascites fluid. Liquid. Contains no preservative.
Unpurified ascites fluid

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
NIH/3T3 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoping Wu et al.
Cell reports, 40(3), 111085-111085 (2022-07-21)
Tuberous sclerosis complex (TSC) is a developmental disorder associated with epilepsy, autism, and cognitive impairment. Despite inactivating mutations in the TSC1 or TSC2 genes and hyperactive mechanistic target of rapamycin (mTOR) signaling, the mechanisms underlying TSC-associated neurological symptoms remain incompletely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service