05-706
Anti-CRMP1 Antibody, clone Y21
ascites fluid, clone Y21, Upstate®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
mouse
Quality Level
antibody form
ascites fluid
antibody product type
primary antibodies
clone
Y21, monoclonal
species reactivity
human
manufacturer/tradename
Upstate®
technique(s)
western blot: suitable
isotype
IgMκ
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... CRMP1(1400)
Specificity
CRMP1
Immunogen
His6-tagged fusion protein corresponding to amino acids 76-572 of human CRMP1
Application
Research Category
Neuroscience
Neuroscience
Research Sub Category
Growth Cones & Axon Guidance
Growth Cones & Axon Guidance
This Anti-CRMP1 Antibody, clone Y21 is validated for use in WB for the detection of CRMP1.
Quality
routinely evaluated by immunoblot on lung adenocarcinoma CL-1 and HeLa whole cell lysates
Target description
~60-65kDa
Physical form
Ascites
mouse ascites with 0.05% sodium azide before the addition of glycerol to 30%
Storage and Stability
2 years at -20°C
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Collapsin response mediator protein-1: a lung cancer invasion suppressor gene with nerve.
Journal of the National Cancer Institute, 93(18), 1364-1365 (2001-09-20)
Journal of the National Cancer Institute, 93(18), 1392-1400 (2001-09-20)
Numerous genetic changes are associated with metastasis and invasion of cancer cells. To identify differentially expressed invasion-associated genes, we screened a panel of lung cancer cell lines (CL(1-0), CL(1-1), CL(1-5), and CL(1-5)-F(4) in order of increasing invasive activity) for such
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service