Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

116811

Sigma-Aldrich

cAMP Direct Immunoassay Kit, Colorimetric

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$616.00

$616.00


Estimated to ship onMarch 23, 2025



Select a Size

Change View
1 KIT
$616.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

$616.00


Estimated to ship onMarch 23, 2025


Quality Level

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Repr. 2 - Skin Corr. 1A - STOT RE 2 Oral - STOT SE 3

target_organs

Kidney,Liver,Blood, Respiratory system

Storage Class

3 - Flammable liquids


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service