1.46715
Fluid Thioglycollate Medium
Tween® 0.5 %, bottle capacity 125 mL, bottle filling volume 100 mL, closure type, Red crimp cap with septum, pack of 10 bottles
Synonym(s):
FTM, Fluid Thioglycollate Medium, Fluid Thioglycollate Medium + 0.5% Tween® 80
About This Item
Recommended Products
agency
EP 2.6.1
JP 4.06
USP 71
71
Quality Level
sterility
sterile; autoclaved
form
liquid
feature
closure type Red crimp cap with septum
ready-to-use
composition
Tween®, 0.5%
packaging
pack of 10 bottles
bottle capacity
125 mL
bottle filling volume
100 mL
capacity
125 mL
pH
7.1 ± 0.2
application(s)
cosmetics
food and beverages
pharmaceutical
sterility testing
storage temp.
2-25°C
suitability
nonselective for
General description
Thioglycollate and L-Cystine in the medium reduces the redox potential of the medium to create an anaerobic atmosphere. Also, it deactivates the presence of mercury and other heavy metals.
The presence of agar further reduces oxygen diffusion and resazurin indicates the reduced oxygen potential in the medium. An increased oxygen concentration will render a color change from yellow to pink in the medium.
Polysorbate 80 inactivates benzyl alcohol, carbanilides, dichlorbenzyl alcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols, phenylethyl alcohols and PHB esters. Histidine is effective against formaldehyde and formaldehyde releasing agents.
application
Fluid Thioglycollate Medium is recommended for sterility testing of samples according to EU and US pharmacopeia.
Features and Benefits
- Our ready-to-use media provide the highest level of quality and testing confidence. They are formulated and tested to meet the pharmacopeia requirements.
- Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests.
- Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, thus improving accuracy, and reducing the risk of incorrect interpretation and false results.
Legal Information
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service