48-691MAG
MILLIPLEX® Human Cell Health Magnetic Bead Panel

Synonym(s):
CH multiplex kit, Human cell health multiplex kit, Luminex CH panel, Luminex human cell health immunoassay, MILLIPLEX® Human Cell Health Magnetic Bead Panel, Millipore cell health panel, cell health multiplex kit
About This Item
Recommended Products
species reactivity
human
Quality Level
manufacturer/tradename
Milliplex®
assay range
intra-assay cv: ≤14%
inter-assay cv: ≤22%
technique(s)
multiplexing: suitable (: Suitable)
input
sample type cell lysate
detection method
fluorometric (Luminex® xMAP®)
storage temp.
2-8°C
General description
Specificity
application
- This is an overnight assay.
- Each assay requires 25 µL per well of diluted or neat stimulated cell lysate.
- This is a qualitative assay and provides insights into the expression levels of the mentioned analytes in response to stress/treatment or condition.
- The suggested working range of protein concentration for the assay is 1 to 25 μg of total protein/well (25 μL/well at 40 to 1000 μg/mL).
- The panel can provide mechanistic insights to researchers performing drug tox screen, studying changes in cell health due to disease/stress, or to study the expression patterns of analytes involved in various cell death and survival pathways.
Features and Benefits
Legal Information
Disclaimer
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service