Skip to Content
Merck
All Photos(1)

Key Documents

6.18048

Supelco

Lithium chloride

anhydrous, purified grade

Synonym(s):

Lithium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
ClLi
CAS Number:
Molecular Weight:
42.39
MDL number:
UNSPSC Code:
12352302
grade:
anhydrous
purified grade

grade

anhydrous
purified grade

SMILES string

[Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service