Skip to Content
Merck
All Photos(1)

Key Documents

AB10130

Sigma-Aldrich

Anti-NR1D1 (Rev-ErbA α) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

V-erbA-related protein EAR-1, nuclear receptor subfamily 1, group D, member 1, thyroid hormone receptor, alpha-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NR1D1(9572)

General description

Rev-ErbA alpha (NR1D1 or nuclear receptor subfamily 1, group D, member 1), is a member of the Rev-ErbA family of nuclear receptors and is a transcriptional repressor. In mammals, Rev-Erbα is highly expressed in the liver, skeletal muscle, adipose tissue, and the brain, participating in the development and circadian regulation of these tissues. Rev-Erbα regulates gene transcription by directly binding to target response elements (RevREs), comprises an A/T-rich flank followed by AGGTCA. Rev-Erbα mediates repression by recruiting the corepressor N-CoR, which then activates the histone deacetylase (HDAC) 3. A number of target genes has been identified for Rev-Erbα, including the lipoproteins ApoA1 and ApoCIII, hydratase dehydrogenase, the circadian factor BMAL, and the anti-fibrinolytic factor PAI-1. Many of these genes are coordinately regulated by Rev-erbα and the RAR-related orphan receptor RORα, which share the same response elements but exert opposite effects on gene transcription. Crosstalk between Rev-Erbα and RORα likely acts to fine-tune of their target physiologic networks, such as circadian rhythms, metabolic homeostasis, and inflammation.

Specificity

Mouse and rat (92% sequence homology).
This antibody recognizes NR1D1 near the N-terminus.

Immunogen

Epitope: Near N-terminus
KLH-conjugated linear peptide corresponding to residues surrounding the area near the N-terminus region of NR1D1.

Application

Anti-NR1D1 (Rev-ErbA α) Antibody is a Rabbit Polyclonal Antibody for detection of NR1D1 (Rev-ErbA α) also known as V-erbA-related protein EAR-1 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blot in A431 cell lysate.
Western Blot Analysis: 1 µg/mL of this antibody detected NR1D1 in 10 µg of A431 cell lysate.

Target description

~67 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-glycine, pH 7.4, 0.150 mM NaCl and 0.05% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C for 1 year from date of receipt.

Analysis Note

Control
A431 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MengLin Chen et al.
Frontiers in pharmacology, 12, 764124-764124 (2021-12-11)
CYP2B10 is responsible for metabolism and detoxification of many clinical drugs. Here, we aimed to investigate a potential role of Period 2 (PER2) in regulating expression of hepatic CYP2B10. Regulatory effects of PER2 on hepatic expression of CYP2B10 and other
Pauline J van der Watt et al.
Molecular cancer research : MCR, 18(9), 1340-1353 (2020-06-07)
There is accumulating evidence for a link between circadian clock disruption and cancer progression. In this study, the circadian clock was investigated in cervical and esophageal cancers, to determine whether it is disrupted in these cancer types. Oncomine datamining revealed
Shogo Sato et al.
Journal of immunology (Baltimore, Md. : 1950), 192(1), 407-417 (2013-12-07)
Disruption of the circadian rhythm is a contributory factor to clinical and pathophysiological conditions, including cancer, the metabolic syndrome, and inflammation. Chronic and systemic inflammation are a potential trigger of type 2 diabetes and cardiovascular disease and are caused by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service