Skip to Content
Merck
All Photos(1)

Key Documents

AB1963

Sigma-Aldrich

Anti-N-WASP Antibody

Chemicon®, from rabbit

Synonym(s):

Neural Wiskott-Aldrich Syndrome Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WASL(8976)

General description

Members of the Wiskott-Aldrich sydrome protein (WASP) family regulate the formation of actin-based cell structures in many cell types. These proteins contain C-terminal actin-binding domains that can stimulate actin polymerization. In addition, these proteins bind the ARP2/3 complex, which can nucleate actin polymerization at sites that lead to branched actin structures. WASP is expressed primarily in hematopoietic cells, while its homolog N-WASP is widely expressed. These proteins have 48% identity in human with the highest homology in the functional regions of these proteins. Serine and tyrosine phosphorylation regulates the activity of both proteins. WASP is observed as a 63 kDa protein in hematopoietic cells, while N-WASP is observed as a 65 kDa in many tissues, especially brain.

Specificity

This antibody detects a 65 kDa protein corresponding to the molecular weight of N-WASP on SDS-PAGE immunoblots of neonatal rat brain lysate. It is also detects 65 kDa proteins in A431, human endothelial, and SKN-SH cells. It does not recognize the 63 kDa WASP protein in Jurkat cell lysate.

Immunogen

Synthetic peptide (coupled to KLH) corresponding to amino acid residues in the N-terminus of human N-WASP. This N-WASP peptide sequence is 100% homologous to rat and mouse N-WASP, and has low homology to the corresponding region in the human WASP sequence.

Application

Anti-N-WASP Antibody is an antibody against N-WASP for use in ELISA & WB.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling
Western blot: 1:1000 ELISA: 1:2000

Physical form

Affinity-purified antibody is supplied in 100 μl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide.

Storage and Stability

Maintain at -20°C for up to one year from date of receipt. Do not aliquot.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

not combustible

flash_point_c

not combustible


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veronica J Jessick et al.
International journal of physiology, pathophysiology and pharmacology, 5(4), 216-227 (2014-01-01)
Neuronal morphology is highly sensitive to ischemia, although some re-organization may promote neuroprotection. In this study we investigate the role of actin regulating proteins (ARP2, ARP3 and WAVE-1) and their role in rapid ischemic tolerance. Using an established in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service