Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABC337

Sigma-Aldrich

Anti-BAFF receptor Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Tumor necrosis factor receptor superfamily member 13C, B-cell-activating factor receptor, BAFF receptor, BAFF-R, BLyS receptor 3, CD268

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

B-cell-activating factor receptor (BAFF receptor, BAFF-R), also known as tumor necrosis factor receptor superfamily member 13C (TNFRSF13C) and BLyS receptor 3 (BR3), is a member of the TNF superfamily that regulate immune responses and induce apoptosis. BAFF receptor promotes the survival of mature B-cells and the B-cell response. BAFF and its receptors are involved in B cell associated autoimmune diseases, and activate NF-kB and c-jun N-terminal kinase. BAFF receptor is highly expressed in spleen and lymph node, with lower levels found in activated B-cells, resting CD4+ T-cells, in thymus and peripheral blood leukocytes. Mutations in BAFF receptor have been associated with immunodeficiency common variable type 4 (CVID4), also called BAFFR defect.

Specificity

This antibody recognizes the C-terminus of BAFF receptor.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of human BAFF receptor.

Application

Detect B-cell-activating factor receptor using this rabbit polyclonal antibody, Anti-BAFF receptor Antibody validated for use in western blotting & IHC.
Immunohistochemistry Analysis: 5 µg/mL from a representative lot detected BAFF receptor in human tonsil tissue.

Quality

Evaluated by Western Blotting in human, mouse, and rat spleen tissue lysates.

Western Blotting Analysis: 5 µg/mL of this antibody detected BAFF receptor in 15 µg of human, mouse, and rat spleen tissue lysates.

Target description

~22 kDa observed. Uniprot describes two isoforms at ~19 kDa Uncharacterized bands may appear in some lysates.

Analysis Note

Control
Human, mouse, and rat spleen tissue lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service