Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABE1847

Sigma-Aldrich

Anti-phospho Mis18-binding protein 1 (Thr702) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Mis18-binding protein 1, phospho Mis18-binding protein 1 (Thr702), Kinetochore null 2 homolog, Kinetochore-associated protein KNL-2 homolog, hsKNL-2, P243, Putative protein p243 which interacts with transcription factor Sp1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pThr702 )

Gene Information

General description

Mis18-binding protein 1 (UniProt Q6P0N0; also known as hsKNL-2, Kinetochore null 2 homolog, Kinetochore-associated protein KNL-2 homolog, P243, Putative protein p243 which interacts with transcription factor Sp1) is encoded by the MIS18BP1 (also known as C14orf106, KIAA1903, KNL2, M18BP1) gene (Gene ID 55320) in human. Mis18-binding protein 1 is a component of the Mis18 complex (Mis18α, Mis18β, and M18BP1/KNL2) that plays a role in mediating cell cycle-dependent deposition/incorporation of the histone H3 variant CENP-A at the centromeres. CENP-A incorporation is carried out by at least two sets of events, the Mis18 complex must first localize to centromeres at the onset of anaphase, HJURP-mediated CENP-A deposition then occurs during G1. Polo-like kinase 1 (Plk1) is shown to phosphorylate M18BP1 on Thr702 and plays an important role in promoting Mis18 complex centromere localization.

Specificity

Specifically reacts with the immunogen peptide, but not the corresponding non-phosphorylated sequence, by dot blot analysis. Phosphorylation site is not present in the spliced isoform 2 of human Mis18-binding protein 1/M18BP1.

Immunogen

Epitope: M18BP1 pThr702
Linear peptide corresponding to a human M18BP1 sequence containing phosphorylated Thr702.

application

Anti-phospho Mis18-binding protein 1 (Thr702) Antibody is an antibody against phospho Mis18-binding protein 1 for use in Immunocytochemistry, Dot Blot, Western Blotting.
Immunocytochemistry Analysis: 4.0 µg/mL from a representative lot detected phospho Mis18-binding protein 1 (Thr702) in HeLa cells.
Dot Blot Analysis: 1 µg/mL from a representative lot detected the immunogen peptide, but not the corresponding non-phosphorylated sequence, by dot blot analysis.
Immunocytochemistry Analysis: A representative lot detected Thr702 phosphorylated Mis18-binding protein 1/M18BP1 localized at the centromeres in HeLa cells (McKinley, K.L., and Cheeseman, I.M. (2014). Cell. 158(2):397-411).
Western Blotting Analysis: A representative lot detected in vitro phosphorylation of recombinant Mis18-binding protein 1/M18BP1 on Thr702 by recombinant polo-like kinase 1 (Plk1) (McKinley, K.L., and Cheeseman, I.M. (2014). Cell. 158(2):397-411).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: 4.0 µg/mL of this antibody detected phospho Mis18-binding protein 1 (Thr702) in A431 cells.

Target description

129 kDa calculated.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide and 30% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service