Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABS2131M

Sigma-Aldrich

Anti-phospho Rab1 (Thr75)

from rabbit

Synonym(s):

YPT-1 related protein, Ras-related protein Rab-1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), mouse (based on 100% sequence homology), bovine (based on 100% sequence homology), sheep (based on 100% sequence homology), rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

phosphorylation (pThr75)

Gene Information

human ... RAB1A(5861)

Related Categories

General description

Ras-related protein Rab-1A (UniProt: P62820; also known as YPT1-related protein) is encoded by the RAB1A (also known as RAB1) gene (Gene ID: 5861) in human. Rab1 is a member of the small GTPase superfamily. Rab proteins are the largest family of small Ras-like GTPases. Three isoforms of Rab1 have been reported that are produced by alternative splicing. Rab GTPases are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. They cycle between an inactive GDP-bound form and an active GTP-bound form, which is able to recruit to membranes different sets of downstream effectors directly responsible for vesicle formation, movement, tethering, and fusion. Rab1 is reported to regulate vesicular protein transport from the endoplasmic reticulum (ER) to the Golgi compartment and on to the cell surface. Rab1 is also shown to be involved in autophagosome assembly and cellular defense reactions against pathogenic bacteria. Rab1 is shown to be phosphorylated at Thr75 by TGF-beta activated kinase 1 (TAK1), which preferentially phosphorylates the inactive (GDP-bound) state of Rab1. Phosphorylation disrupts interaction with GDP dissociation inhibitor 1 (GDI1), but not GEF or GAP. Phosphorylation at Thr75 is shown to be essential for Rab1 function.

Specificity

This rabbit polyclonal antibody detects Ras-related protein Rab-1A (Rab1) in human cells. It targets an epitope within 14 amino acids surrounding phosphorylated Thr75.

Immunogen

A linear peptide corresponding to 14 amino acids surrounding phosphorylated Thr75 from the N-terminal half of human Ras-related protein Rab-1A.
Epitope: unknown

Application

Anti-phospho Rab1 (Thr75), Cat. No. ABS2131, is a rabbit polyclonal antibody that detects Ras-related protein Rab-1A and has been tested for use in Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A representative lot detected phospho Rab1 (Thr75) in Western Blotting applications (Levin, R.S., et. al. (2016). Proc Natl Acad Sci USA. 113(33):E4776-83).

Quality

Evaluated by Western Blotting in lysate from HEK293T stably expressing GST-Rab1a-Tak1.


Western Blotting Analysis: A 1:250 dilution of this antibody detected phospho Rab1 (Thr75) in lysates from HEK-293T stably expressing GST-Rab1a-Tak1.

Target description

~48 kDa observed; 22.68 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in PBS with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service