EZFREAD01US
EZ-Fluo™ Reader
Contains EZ-Fluo™ reader, power cord for United States
Synonym(s):
EZ-Fluo™ Reader, For rapid detection of microbial contamination in filterable samples, suitable for bioburden testing, Rapid Microbial Detection System
About This Item
AC/DC input
100 V / 240 V, 50-60 Hz
material
304L stainless steel (optical chamber)
304L stainless steel handle
aluminum housing (sheet)
polyester foot
polyester screen
agency
EP
manufacturer/tradename
EZ-Fluo™
availability
available only in USA
parameter
15-40 °C temperature
4.4 kg operating weight
L × W × H
9.8 in. × 5.6 × 4.9
application(s)
environmental
food and beverages
compatibility
for use with EZ-Fluo™ Rapid Detection System.
Looking for similar products? Visit Product Comparison Guide
General description
The method used is fast, non-destructive and fluorescent staining based. The non-destructive feature allows the user to re-incubate the filtered sample for growth on non-selective microbial media.
Application
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service