MABF1775
Mouse IgG2a Negative Control, clone C1.18.4, APC conjugate Antibody
clone C1.18.4, from mouse, purified by affinity chromatography
Synonym(s):
Immunoglobulin G 2a, IgG2a
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
clone
C1.18.4, monoclonal
purified by
affinity chromatography
technique(s)
flow cytometry: suitable
isotype
IgG2a
shipped in
wet ice
target post-translational modification
unmodified
General description
The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.
Immunogen
Immunogen information for this antibody is proprietary information.
Application
Research Category
Inflammation & Immunology
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
Immunoglobulins & Immunology
The isotype control should be used at the same concentration as the primary antibody for which it is a control.
This mouse monoclonal Mouse IgG2a Negative Control, clone C1.18.4, APC conjugate, Cat. No. MABF1775 is validated for use in Flow Cytometry.
Physical form
Purified Mouse monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2. This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography.
Purified by size-exclusion chromatography
Storage and Stability
Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service