MABF2312
Anti-CD11a Antibody, clone TS2/4
Synonym(s):
CD11 antigen-like family member A, Integrin alpha-L, LFA-1A, Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain
About This Item
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
TS2/4, monoclonal
mol wt
calculated mol wt 128.770 kDa
observed mol wt ~N/A kDa
purified by
using protein G
species reactivity
human
packaging
antibody small pack of 100
technique(s)
affinity chromatography: suitable
electrophoresis: suitable
flow cytometry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
inhibition assay: suitable
radioimmunoassay: suitable
isotype
IgG1κ
epitope sequence
Extracellular domain
Protein ID accession no.
UniProt accession no.
storage temp.
-10 to -25°C
Gene Information
human ... ITGAL(3683)
Specificity
Immunogen
Application
Evaluated by Flow Cytometry in THP-1 cells.
Flow Cytometry Analysis (FC): 1.0 µg of this antibody detected CD11a/ITGAL in one million THP-1 cells.
Tested Applications
Immunoprecipitation (IP) Application: A representative lot immunoprecipitated CD11a in immunoprecipitation application (Sanchez-Madrid, F., et al. (1982). Proc. Natl. Acad. Sci. USA. 79(23); 7489-7493).
Gel Electrophoresis Application: A representative lot detected CD11a in Gel Electrophoresis application (Sanchez-Madrid, F., et al. (1982). Proc. Natl. Acad. Sci. USA. 79(23); 7489-7493).
Inhibition Assay: A representative lot inhibited CTL-mediated killing of Daudi cells. (Sanchez-Madrid, F., et al. (1982). Proc. Natl. Acad. Sci. USA. 79(23); 7489-7493).
Radioimmunoassay Application: A representative lot detected CD11a in Radioimmunoassay applications (Dustin, ML., et al. (1992). J. Immunol. 148(9); 2654-2663).
Immunoaffinity Purification: A representative lot was used for immunoaffinity purification of CD11a. (Dustin, ML., et al. (1992). J. Immunol. 148(9); 2654-2663).
Immunofluorescence Application: A representative lot detected CD11a in Immunofluorescence microscopy application. (Dustin, ML., et al. (1992). J. Immunol. 148(9); 2654-2663).
Flow Cytometry (FC) Application: A representative lot detected CD11a in Flow Cytometry application. (Sanchez-Madrid, F., et al. (1982). Proc Natl Acad Sci USA. 79(23):7489-93; Samuelsson, M., et al. (2017). Sci Signal. 10(509):eaai8629.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service