MABF2751
Anti-Murine Coronavirus MHV-JHM nucleocapsid Antibody, clone 1-16-1
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
1-16-1, monoclonal
mol wt
calculated mol wt 49.73 kDa
observed mol wt ~50 kDa
purified by
using protein G
species reactivity
SARS coronavirus
packaging
antibody small pack of 100 μL
technique(s)
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG1κ
epitope sequence
Unknown
Protein ID accession no.
UniProt accession no.
shipped in
2-8°C
target post-translational modification
unmodified
Gene Information
vaccinia virus ... n> N(1489757)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in Mouse delayed brain tumor (DBT) cell lysates.
Western Blotting Analysis: A 1:500 dilution of this antibody detected Murine Coronavirus MHV-JHM nucleocapsid in Mouse delayed brain tumor (DBT) cell lysates.
Tested Applications
Western Blotting Analysis: A representative lot detected Murine Coronavirus MHV-JHM nucleocapsid in Western Blotting applications (Leibowitz, J.L., et al. (1987). Adv Exp Med Biol. 218; 321-31).
Immunofluorescence Analysis: A representative lot detected Murine Coronavirus MHV-JHM nucleocapsid in Immunofluorescence applications (Nanda, S.K., et al. (2001). J Virol. 75(7):3352-62).
Immunohistochemistry Applications: A representative lot detected Murine Coronavirus MHV-JHM nucleocapsid in Immunohistochemistry applications (Miura, T.A., et al. (2008). J Virol. 82(2):755-63; Navas, S., et al. (2001). J Virol. 75(5):2452-7).
Immunoprecipitation Analysis: A representative lot detected Murine Coronavirus MHV-JHM nucleocapsid in Immunoprecipitation applications (Nanda, S.K., et al. (2004). Arch Virol. 149(1):93-111).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service