Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT1539

Sigma-Aldrich

Anti-Filaggrin Antibody, clone AHF3

Synonym(s):

FLG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
clone:
AHF3, monoclonal
application:
ELISA
EM
ICC
IF
IHC
WB
species reactivity:
human
technique(s):
ELISA: suitable
electron microscopy: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable
citations:

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

AHF3, monoclonal

mol wt

calculated mol wt 435.17 kDa
observed mol wt ~N/A kDa

purified by

ion-exchange chromatography

species reactivity

human

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
electron microscopy: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

Gene Information

human ... FLG(2312)

Specificity

Clone AHF3 is a mouse monoclonal antibody that detects Filaggrin and Profilaggrin.

Immunogen

Partially purified Filaggrin from human breast skin tissue.

Application

Quality Control Testing

Evaluated by Immunocytochemistry in HaCat cells.

Immunocytochemistry Analysis: A 1:1,000 dilution of this antibody detected Filaggrin in HaCat cells.

Tested Applications

Immunohistochemistry Applications: A representative lot detected Filaggrin in Immunohistochemistry application.. (Henry, J., et al. (2011). FASEB J. 25(5):1567-76; Hsu, C-Y., et al. (2011). J Biol Chem. 286(26):23222-33).

Western Blotting Analysis: A representative lot detected Filaggrin in Western Blotting application. (Simon, M., et al. (1995). J Invest Dermatol. 105(3):432-7; Simon, M., et al. (1996). Biochem J. 317(Pt1)(Pt1):173-7; Henry, J., et al. (2011). FASEB J. 25(5):1567-76; Hsu, C-Y., et al. (2011). J Biol Chem. 286(26):23222-33; Pellerin, L., et al. (2013). J Allergy Clin Immunol. 131(4):1094-102; Hsu, C-Y., et al. (2017). J Invest Dermatol. 137(2):422-429).

Electron Microscopy: A representative lot detected Filaggrin in Electron Microscopy application. (Simon, M., et al. (1995). J Invest Dermatol. 105(3):432-7; Henry, J., et al. (2011). FASEB J. 25(5):1567-76).

Immunofluorescence Analysis: A representative lot detected Filaggrin in Immunofluorscence application. (Simon, M., et al. (1995). J Invest Dermatol. 105(3):432-7; Henry, J., et al. (2011). FASEB J. 25(5):1567-76; Hsu, C-Y., et al. (2011). J Biol Chem. 286(26):23222-33).

ELISA Analysis: A representative lot detected Filaggrin in ELISA application. (Simon, M., et al. (1995). Clin Exp Immunol. 100(1):90-8; Simon, M., et al. (1996). Biochem J. 317(Pt1)(Pt1):173-7; Miedouge, M., et al. (2001). Tumour Biol. 22(1):19-26).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Filaggrin (UniProt: P20930; also known as FLG) is encoded by the FLG gene (Gene ID: 2312) in human. Filaggrin, a calcium-binding protein, is synthesized as a large, insoluble, highly phosphorylated precursor with several tandem copies of 324 amino acids in cytoplasmic granules. During terminal differentiation it undergoes dephosphorylation and proteolytic cleavage to remove linker peptides and produce functional basic filaggrin units. It is widely expressed and in skin tissue it is detected mainly in stratum granulosum of the epidermis. During terminal differentiation of mammalian epidermis, it aggregates keratin intermediate filaments and promotes disulfide-bond formation among the intermediate filaments. This creates the intracellular fibrous matrix of the cornified cells. Deimination of arginine to citrulline by peptidylarginine deiminase is reported to result in reduction in its affinity for cytokeratins. The N-terminal region of the mature filaggrin is shown to be heterogenous and is blocked by the formation of pyroglutamate. Clone AHF3 detects both the profilaggrin and filaggrin and exhibits reactivity in various immunochemical applications. (Ref.: Simon, M., et al. (1996). Biochem. J. 317(pt 1); 173 177; Simon, M., et al. (1995). Clin. Exp. Immunol. 100(1); 90-98; Simon, M., et al. (1995). J. Invest. Dermatol. 105(3); 432-437).

Physical form

Purified mouse monoclonal antibodyIgG1κ in buffer containing PBS with 0.1% sodium azide with 50% glycerol.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service