MANTLTKIT
MAS-100 NT® Validation Protocol and Test Record Protocol kit
Letter format
All Photos(1)
About This Item
UNSPSC Code:
41116500
compatibility
for use with MAS-100 NT® air sampler system.
General description
MAS-100 NT® validation protocol and associated test record protocol kit is designed as a ready to use guide to perform the validation activities for the MAS-100 NT® air sampling pump. The validation protocol kit ensures regulatory compliance of the pumps and guarantees consistent performance.
Application
MAS-100 NT® validation protocol kit is used for the validation of MAS-100 NT® pumps which samples environmental air inside cleanrooms and isolators.
Legal Information
MAS-100 NT is a registered trademark of MBV AG