NW-10X5ML-U
Unlabeled Vials Reagent - Rare Specificities & AHG
BIOSCOT™ Anti-Fya from the clone P3TIM, Human IgG Antibody, Blood Grouping Reagent
Synonym(s):
Anti-Fya (clone P3TIM), Blood Grouping Reagent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116102
NACRES:
NA.41
Note: For further information on price, availability or to place an order, please contact us
biological source
human
Quality Level
antibody form
unpurified
clone
P3TIM, monoclonal
form
liquid
packaging
pkg of 10 × 5 mL
concentration
2-6 mg/mL protein (Total Protein Range)
technique(s)
agglutination assay: suitable
pH
6.8-7.8
isotype
IgG
Related Categories
General description
Contains 0.1% (w/v) Sodium Azide
Filtered through a 0.22μm terminal filter
Shelf Life: 36 months
Product formulation conforms to the requirements of the EU IVD directive.
For In Vitro use only
Filtered through a 0.22μm terminal filter
Shelf Life: 36 months
Product formulation conforms to the requirements of the EU IVD directive.
For In Vitro use only
Application
For use by Tube;Gel and Column techniques
Vialled, Unlabeled
Legal Information
BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service