Key Documents
SPKGENINS028
Millipore
Spare part, Electromagnetic Flowmeter
1/4in Qmax 3 L/min
About This Item
UNSPSC Code:
23151800
Pricing and availability is not currently available.
material
polysulfone
Quality Level
product line
Mobius®
compatibility
for use with Mobius®
General description
This electromagnetic flowmeter mesures the flow rate of conductive liquids thanks to a magnetic field.
This spare part is classified as ′Critical,′ meaning that it is crucial, for avoiding system downtime, to keep at least one in stock for immediate availability.
Packaging
Pack of 1 unit
Legal Information
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany