Skip to Content
Merck
All Photos(1)

Key Documents

ZWACIDC12

ROCare A

new

Acid cleaning chemical removes scaling from RO membranes

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41104210

Quality Level

General description

ROCare A is an acidic cleaner that protects the water purification systems by effectively removing some or most mineral scale buildup on the surface of the reverse osmosis (RO) membranes. It is formulated with citric acid only, significantly reducing toxicity, ensuring safety and supporting environmental sustainability.

The acid cleaner is delivered in a pouch and the empty pouch is removed after use. It is to be used periodically when prompted by the system, or when the RO % rejection drops more than 5%, or when the RO permeate flow rate drops more than 10% due to mineral scaling.

The ROCare A pouch cleans RO membranes supporting Milli-Q® HR/HX/CLX, Elix® Large, Milli-Q® IX, Milli-Q® IQ, Milli-Q® EQ, Elix® Advantage, Milli-Q® Integral and Essential water purification systems.

Application

Removes scaling from RO membranes.

Features and Benefits

  • Anti-scaling agent
  • Formulated with citric acid only, to reduce toxicity, ensure safety and support environment sustainability
  • Contains 12 packages

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service