28-3640
Sodium perchlorate monohydrate
SAJ first grade, ≥97.0%
About This Item
Recommended Products
grade
SAJ first grade
assay
≥97.0%
form
crystalline
reaction suitability
reagent type: oxidant
availability
available only in Japan
pH
4.5-7.0 (25 °C, 50 g/L)
SMILES string
O.[Na+].[O-]Cl(=O)(=O)=O
InChI
1S/ClHO4.Na.H2O/c2-1(3,4)5;;/h(H,2,3,4,5);;1H2/q;+1;/p-1
InChI key
IXGNPUSUVRTQGW-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 1
Storage Class
5.1A - Strongly oxidizing hazardous materials
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service