Skip to Content
Merck
All Photos(1)

Key Documents

03077

Millipore

Liver Hydrolysate

suitable for microbiology

Synonym(s):

Peptone from liver

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41106212
NACRES:
NA.85

Pricing and availability is not currently available.

biological source

Porcine liver

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

amino-nitrogen, ≥4%
total nitrogen (N), ≥10%

ign. residue

≤18%

loss

≤6% loss on drying

pH

6.9±0.5 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear, brownish-yellow to brown

application(s)

food and beverages
microbiology

Looking for similar products? Visit Product Comparison Guide

General description

A biologically standardised enzymatic digest of fresh swine liver followed by a careful desiccation process to maintain its fundamental characteristics. For use as a source of nutrients in microbiological culture media.The digest is water soluble and compatible with other culture media ingredients. Being derived from liver this product contains relatively high levels of iron. Liver peptone is employed in culture media for Trichomonas and other fastidious protozoa, and for some pathogen and saprofitic fungi, microplasms and anaerobic bacteria.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service